Product Details

SNP ID
rs202172957
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30429967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGTGTCCCCACGAATCTCCCCA[A/C]CGGCCACAGACATGCCGCCCACCGC
Phenotype
MIM: 615840
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCTPP1 PubMed Links

Gene Details

Gene
DCTPP1
Gene Name
dCTP pyrophosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024096.1 86 Missense Mutation GGT,GTT G5V NP_077001.1

View Full Product Details