Product Details

SNP ID
rs200242137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63819983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTTTCTCCTTGCCAAGCCCAGCC[G/T]TCTTGTAGAGACTACAGGGGGGAAG
Phenotype
MIM: 613369 MIM: 601681
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DDX42 PubMed Links

Gene Details

Gene
DDX42
Gene Name
DEAD-box helicase 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007372.3 3009 Intron NP_031398.2
NM_203499.2 3009 Intron NP_987095.1
XM_006721657.1 3009 Intron XP_006721720.1
XM_011524259.2 3009 Intron XP_011522561.1
XM_017024111.1 3009 Intron XP_016879600.1
XM_017024112.1 3009 Intron XP_016879601.1
Gene
FTSJ3
Gene Name
FtsJ homolog 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017647.3 3009 Missense Mutation AAG,ACG K788T NP_060117.3
Gene
PSMC5
Gene Name
proteasome 26S subunit, ATPase 5
There are no transcripts associated with this gene.

View Full Product Details