Product Details

SNP ID
rs199533241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28319346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGACCTGCAGGCGGTGCTGCCGC[G/T]CGAGCTCTACCCAGTCGAGGTGAGG
Phenotype
MIM: 614394 MIM: 612912
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IFT20 PubMed Links

Gene Details

Gene
IFT20
Gene Name
intraflagellar transport 20
There are no transcripts associated with this gene.

Gene
TMEM97
Gene Name
transmembrane protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014573.2 306 Missense Mutation CGC,CTC R36L NP_055388.2
XM_005257965.3 306 Missense Mutation GCG,TCG A77S XP_005258022.1

View Full Product Details