Product Details

SNP ID
rs199914191
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:413385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGAATTCTTGCTGACCCGGAGG[A/C]AGAGAAGGAAAGGATTCGCATCTAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C17orf97 PubMed Links

Gene Details

Gene
C17orf97
Gene Name
chromosome 17 open reading frame 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013672.4 558 Missense Mutation GAA,GCA E181A NP_001013694.4
Gene
LOC105371430
Gene Name
uncharacterized LOC105371430
There are no transcripts associated with this gene.

View Full Product Details