Product Details
- SNP ID
-
rs200055352
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:10301698 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTGGACAGCTGCGTGTTGGCCTG[C/T]TCCTCCTGCACAGGAGACAGAGGGG
- Phenotype
-
MIM: 603487
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYH13
PubMed Links
Gene Details
- Gene
- MYH13
- Gene Name
- myosin heavy chain 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003802.2 |
5763 |
Silent Mutation |
GAA,GAG |
E1891E |
NP_003793.2 |
View Full Product Details