Product Details
- SNP ID
-
rs200224909
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40660187 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAATGCTGCTCGCTGGCATGTAG[C/G]GAGTTTTCAAGGCCCCTTTCCTGTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT222
PubMed Links
Gene Details
- Gene
- KRT222
- Gene Name
- keratin 222
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152349.2 |
311 |
Silent Mutation |
TCC,TCG |
S82S |
NP_689562.1 |
View Full Product Details