Product Details

SNP ID
rs199689510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2057929 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTATGGCGAGCCCGAGAGCTT[C/T]GGTGACAACCTGTACGTGTGCATTC
Phenotype
MIM: 603825 MIM: 610016 MIM: 613487 MIM: 610963
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIC1 PubMed Links

Gene Details

Gene
HIC1
Gene Name
hypermethylated in cancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098202.1 1296 Silent Mutation TTC,TTT F432F NP_001091672.1
NM_006497.3 1296 Silent Mutation TTC,TTT F413F NP_006488.2
Gene
MIR132
Gene Name
microRNA 132
There are no transcripts associated with this gene.

Gene
MIR212
Gene Name
microRNA 212
There are no transcripts associated with this gene.

Gene
SMG6
Gene Name
SMG6, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details