Product Details

SNP ID
rs199520559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7709360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGCCACCCTGGTCCTGGCTCCTT[C/T]GGGAGGGGAGGGTCTCTGGGCCTGG
Phenotype
MIM: 603333 MIM: 602297 MIM: 612661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAH2 PubMed Links

Gene Details

Gene
DNAH2
Gene Name
dynein axonemal heavy chain 2
There are no transcripts associated with this gene.

Gene
EFNB3
Gene Name
ephrin B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001406.3 1204 Silent Mutation TTC,TTT F269F NP_001397.1
Gene
WRAP53
Gene Name
WD repeat containing antisense to TP53
There are no transcripts associated with this gene.

View Full Product Details