Product Details

SNP ID
rs199762429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36537865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATAACACCACTGCTGTTGATTA[C/T]TTTTCCCCTAAATAAGTCCTGGATT
Phenotype
MIM: 612275 MIM: 610275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GGNBP2 PubMed Links

Gene Details

Gene
GGNBP2
Gene Name
gametogenetin binding protein 2
There are no transcripts associated with this gene.

Gene
MYO19
Gene Name
myosin XIX
There are no transcripts associated with this gene.

Gene
PIGW
Gene Name
phosphatidylinositol glycan anchor biosynthesis class W
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178517.3 1063 Missense Mutation ACT,ATT T255I NP_848612.2
XM_005257238.2 1063 Missense Mutation ACT,ATT T255I XP_005257295.1
XM_011524646.2 1063 Missense Mutation ACT,ATT T255I XP_011522948.1

View Full Product Details