Product Details

SNP ID
rs200037144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40554818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGATGTGCCGACAGGAAGACCAC[A/T]GCCGGAGCTGCTCCTGGCTGAGGCA
Phenotype
MIM: 600242
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCR7 PubMed Links

Gene Details

Gene
CCR7
Gene Name
C-C motif chemokine receptor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301714.1 1142 Missense Mutation CAG,CTG Q291L NP_001288643.1
NM_001301716.1 1142 Missense Mutation CAG,CTG Q348L NP_001288645.1
NM_001301717.1 1142 Missense Mutation CAG,CTG Q348L NP_001288646.1
NM_001301718.1 1142 Missense Mutation CAG,CTG Q348L NP_001288647.1
NM_001838.3 1142 Missense Mutation CAG,CTG Q354L NP_001829.1

View Full Product Details