Product Details

SNP ID
rs202053376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2690678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGCCCCCGCTGGCGCGGGCTC[A/G]GTAGCCATGGGCTCCTCGGCTCTAT
Phenotype
MIM: 616184 MIM: 601545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLUH PubMed Links

Gene Details

Gene
CLUH
Gene Name
clustered mitochondria homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015229.3 3863 Silent Mutation NP_056044.3
XM_011523768.2 3863 Silent Mutation XP_011522070.1
Gene
MIR6776
Gene Name
microRNA 6776
There are no transcripts associated with this gene.

Gene
PAFAH1B1
Gene Name
platelet activating factor acetylhydrolase 1b regulatory subunit 1
There are no transcripts associated with this gene.

View Full Product Details