Product Details

SNP ID
rs201008283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31852176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGTCAAACTGTCCTGTTTCAAAG[A/G]CCATCTTGGAATAATAGGGTATCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPRS PubMed Links

Gene Details

Gene
COPRS
Gene Name
coordinator of PRMT5 and differentiation stimulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018405.3 626 Missense Mutation GCC,GTC A173V NP_060875.2
XM_005258000.1 626 Missense Mutation GCC,GTC A161V XP_005258057.1

View Full Product Details