Product Details

SNP ID
rs201111716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:34932485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGCCACTTCAATTGCACCAGCT[C/T]CAGGAACCATACAACCTATTGGAGA
Phenotype
MIM: 610730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCT6B PubMed Links

Gene Details

Gene
CCT6B
Gene Name
chaperonin containing TCP1 subunit 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193529.2 1210 Missense Mutation GAA,GGA E373G NP_001180458.1
NM_001193530.1 1210 Missense Mutation GAA,GGA E365G NP_001180459.1
NM_006584.3 1210 Missense Mutation GAA,GGA E410G NP_006575.2
XM_011524203.2 1210 Missense Mutation GAA,GGA E268G XP_011522505.1
XM_017024023.1 1210 Missense Mutation GAA,GGA E344G XP_016879512.1
XM_017024024.1 1210 Missense Mutation GAA,GGA E231G XP_016879513.1

View Full Product Details