Product Details

SNP ID
rs201316706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74842498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGGTCCCGGGAAGCCTTGCGT[C/G]CCACGGGCTACCCTGCTGATCTCGT
Phenotype
MIM: 138254
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GRIN2C PubMed Links

Gene Details

Gene
GRIN2C
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000835.4 5086 Silent Mutation GGC,GGG G1213G NP_000826.2
NM_001278553.1 5086 Intron NP_001265482.1
XM_006721845.3 5086 Silent Mutation GGC,GGG G1288G XP_006721908.1
XM_006721846.3 5086 Silent Mutation GGC,GGG G1287G XP_006721909.1
XM_011524686.2 5086 Silent Mutation GGC,GGG G1316G XP_011522988.1
XM_011524687.2 5086 Silent Mutation GGC,GGG G1309G XP_011522989.1
XM_011524688.2 5086 Silent Mutation GGC,GGG G1296G XP_011522990.1
XM_011524689.2 5086 Silent Mutation GGC,GGG G1242G XP_011522991.1
XM_011524692.2 5086 Intron XP_011522994.1
XM_017024544.1 5086 Silent Mutation GGC,GGG G1317G XP_016880033.1
Gene
TMEM104
Gene Name
transmembrane protein 104
There are no transcripts associated with this gene.

View Full Product Details