Product Details

SNP ID
rs201968689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31852159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTCAATCTTCAGCATCGTCAA[A/G]CTGTCCTGTTTCAAAGACCATCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPRS PubMed Links

Gene Details

Gene
COPRS
Gene Name
coordinator of PRMT5 and differentiation stimulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018405.3 643 Missense Mutation CTT,TTT L179F NP_060875.2
XM_005258000.1 643 Missense Mutation CTT,TTT L167F XP_005258057.1

View Full Product Details