Product Details

SNP ID
rs200875679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41724730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCACGACGATCTGCAGCTTGTCC[A/G]GCTGGCGGCAGGGAGGGCCCCGTGG
Phenotype
MIM: 137250 MIM: 600947
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAST PubMed Links

Gene Details

Gene
GAST
Gene Name
gastrin
There are no transcripts associated with this gene.

Gene
HAP1
Gene Name
huntingtin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079870.1 1768 Missense Mutation CGG,TGG R594W NP_001073339.1
NM_001079871.1 1768 Missense Mutation CGG,TGG R586W NP_001073340.1
NM_177977.2 1768 Missense Mutation CGG,TGG R611W NP_817084.2

View Full Product Details