Product Details

SNP ID
rs202042736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48614363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCTCCTCGCCCAGGCCGCTGGC[A/G]GCGGCAAGCTTGCAGTCTGCGTACT
Phenotype
MIM: 142961 MIM: 142962 MIM: 142963 MIM: 142964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXB-AS3 PubMed Links

Gene Details

Gene
HOXB-AS3
Gene Name
HOXB cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXB6
Gene Name
homeobox B6
There are no transcripts associated with this gene.

Gene
HOXB7
Gene Name
homeobox B7
There are no transcripts associated with this gene.

Gene
HOXB8
Gene Name
homeobox B8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024016.3 5893 Silent Mutation GCC,GCT A114A NP_076921.1
XM_005257286.2 5893 Silent Mutation GCC,GCT A114A XP_005257343.1
XM_017024564.1 5893 Silent Mutation GCC,GCT A114A XP_016880053.1
Gene
HOXB9
Gene Name
homeobox B9
There are no transcripts associated with this gene.

View Full Product Details