Product Details

SNP ID
rs202111769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:5130657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTGAGCCCGTTGGAATCAACA[C/G]CAGCATTGATCGTTTTGGCATTTTG
Phenotype
MIM: 604334 MIM: 616463
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP6 PubMed Links

Gene Details

Gene
USP6
Gene Name
ubiquitin specific peptidase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304284.1 2576 Missense Mutation ACC,AGC T43S NP_001291213.1
NM_004505.3 2576 Intron NP_004496.2
XM_011524050.1 2576 Missense Mutation ACC,AGC T43S XP_011522352.1
XM_011524051.2 2576 Missense Mutation ACC,AGC T43S XP_011522353.1
XM_011524052.2 2576 Intron XP_011522354.1
XM_011524053.2 2576 Intron XP_011522355.1
XM_011524054.2 2576 Missense Mutation ACC,AGC T43S XP_011522356.1
XM_011524055.2 2576 Missense Mutation ACC,AGC T43S XP_011522357.1
XM_011524056.2 2576 Intron XP_011522358.1
XM_011524057.2 2576 Missense Mutation ACC,AGC T43S XP_011522359.1
XM_011524058.2 2576 Missense Mutation ACC,AGC T43S XP_011522360.1
XM_011524059.2 2576 Missense Mutation ACC,AGC T43S XP_011522361.1
XM_017025290.1 2576 Missense Mutation ACC,AGC T43S XP_016880779.1
Gene
ZNF232
Gene Name
zinc finger protein 232
There are no transcripts associated with this gene.

View Full Product Details