Product Details

SNP ID
rs200949415
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3216258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCTCTGTGCCATTACTATGCA[C/T]CATTGTCTCCTATATTCGAGTCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1A1 PubMed Links

Gene Details

Gene
OR1A1
Gene Name
olfactory receptor family 1 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014565.2 638 Missense Mutation ACC,ATC T213I NP_055380.2

View Full Product Details