Product Details

SNP ID
rs201018196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78855824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTCCATCCAGAGGCACTCGTCC[A/G]GGGAGGAGATGTAGCACGGGATCAT
Phenotype
MIM: 188825
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TIMP2 PubMed Links

Gene Details

Gene
TIMP2
Gene Name
TIMP metallopeptidase inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003255.4 808 Missense Mutation CCG,CTG P169L NP_003246.1

View Full Product Details