Product Details

SNP ID
rs200915238
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74588616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTTTTGAGATTCTCCATGGTCA[C/T]GGTGAACACGTGGTTTTTCTGATTG
Phenotype
MIM: 616301
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C17orf77 PubMed Links
Additional Information
For this assay, SNP(s) [rs783239] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C17orf77
Gene Name
chromosome 17 open reading frame 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302809.1 303 Intron NP_001289738.1
NM_152460.2 303 Intron NP_689673.2
Gene
CD300LD
Gene Name
CD300 molecule like family member d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001115152.1 303 Missense Mutation ATG,GTG M92V NP_001108624.1
Gene
LOC101928343
Gene Name
uncharacterized LOC101928343
There are no transcripts associated with this gene.

View Full Product Details