Product Details

SNP ID
rs201603103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7630202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCAGAGGCCCACTGGCTACCTC[A/G]CGCCTGCTGCTGTTGCTGCTGTTGC
Phenotype
MIM: 611463 MIM: 182205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAT2 PubMed Links

Gene Details

Gene
SAT2
Gene Name
spermidine/spermine N1-acetyltransferase family member 2
There are no transcripts associated with this gene.

Gene
SHBG
Gene Name
sex hormone binding globulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040.4 109 Silent Mutation TCA,TCG S10S NP_001031.2
NM_001146279.2 109 Silent Mutation TCA,TCG S10S NP_001139751.1
NM_001146280.2 109 Silent Mutation TCA,TCG S10S NP_001139752.1
NM_001146281.2 109 Silent Mutation TCA,TCG S10S NP_001139753.1
NM_001289113.1 109 Intron NP_001276042.1
NM_001289114.1 109 Intron NP_001276043.1
NM_001289115.1 109 Intron NP_001276044.1
NM_001289116.1 109 UTR 5 NP_001276045.1

View Full Product Details