Product Details

SNP ID
rs201160126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3664702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTTGTCCGTCTTGTCTTCAGAGA[A/T]GGGATTCTGGGAAGGATCCTGGTCG
Phenotype
MIM: 606272 MIM: 602836 MIM: 616484
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CTNS PubMed Links

Gene Details

Gene
CTNS
Gene Name
cystinosin, lysosomal cystine transporter
There are no transcripts associated with this gene.

Gene
EMC6
Gene Name
ER membrane protein complex subunit 6
There are no transcripts associated with this gene.

Gene
P2RX5
Gene Name
purinergic receptor P2X 5
There are no transcripts associated with this gene.

Gene
P2RX5-TAX1BP3
Gene Name
P2RX5-TAX1BP3 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TAX1BP3
Gene Name
Tax1 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204698.1 289 Missense Mutation ATC,TTC I46F NP_001191627.1
NM_014604.3 289 Missense Mutation ATC,TTC I46F NP_055419.1

View Full Product Details