Product Details
- SNP ID
-
rs201498132
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:30484705 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAATGGCAGAATATACCAACAGT[A/G]CAGGTGTCCCCTCCTTGAATGCAGC
- Phenotype
-
MIM: 604026
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GOSR1
PubMed Links
Gene Details
- Gene
- GOSR1
- Gene Name
- golgi SNAP receptor complex member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007024.1 |
550 |
Missense Mutation |
ACA,GCA |
T30A |
NP_001007025.1 |
NM_001007025.1 |
550 |
Missense Mutation |
ACA,GCA |
T93A |
NP_001007026.1 |
NM_004871.2 |
550 |
Missense Mutation |
ACA,GCA |
T95A |
NP_004862.1 |
XM_005258070.4 |
550 |
Missense Mutation |
ACA,GCA |
T145A |
XP_005258127.1 |
XM_005258071.4 |
550 |
Missense Mutation |
ACA,GCA |
T143A |
XP_005258128.1 |
XM_005258072.2 |
550 |
Missense Mutation |
ACA,GCA |
T30A |
XP_005258129.1 |
XM_017025374.1 |
550 |
Missense Mutation |
ACA,GCA |
T145A |
XP_016880863.1 |
XM_017025375.1 |
550 |
Missense Mutation |
ACA,GCA |
T95A |
XP_016880864.1 |
XM_017025376.1 |
550 |
Missense Mutation |
ACA,GCA |
T93A |
XP_016880865.1 |
XM_017025377.1 |
550 |
Missense Mutation |
ACA,GCA |
T145A |
XP_016880866.1 |
View Full Product Details