Product Details

SNP ID
rs201330544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7024733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCAGCAGCAGCAGCAGCAGTG[A/C]AGAAGGACCCATTCCTGGTCCCCAG
Phenotype
MIM: 608992 MIM: 610718 MIM: 615672
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BCL6B PubMed Links

Gene Details

Gene
BCL6B
Gene Name
B-cell CLL/lymphoma 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181844.3 799 Missense Mutation GAA,GCA E246A NP_862827.1
Gene
C17orf49
Gene Name
chromosome 17 open reading frame 49
There are no transcripts associated with this gene.

Gene
MIR195
Gene Name
microRNA 195
There are no transcripts associated with this gene.

Gene
MIR497
Gene Name
microRNA 497
There are no transcripts associated with this gene.

Gene
MIR497HG
Gene Name
mir-497-195 cluster host gene
There are no transcripts associated with this gene.

Gene
RNASEK-C17orf49
Gene Name
RNASEK-C17orf49 readthrough
There are no transcripts associated with this gene.

View Full Product Details