Product Details

SNP ID
rs201879172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28319343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATGGACCTGCAGGCGGTGCTGC[C/T]GCGCGAGCTCTACCCAGTCGAGGTG
Phenotype
MIM: 614394 MIM: 612912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT20 PubMed Links

Gene Details

Gene
IFT20
Gene Name
intraflagellar transport 20
There are no transcripts associated with this gene.

Gene
TMEM97
Gene Name
transmembrane protein 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014573.2 303 Missense Mutation CCG,CTG P35L NP_055388.2
XM_005257965.3 303 Missense Mutation CGC,TGC R76C XP_005258022.1

View Full Product Details