Product Details

SNP ID
rs200256118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35369507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGAAGGGTTTCCTTGTCTCTTT[G/T]CAAACCTACCATCTTGTTCATAGGT
Phenotype
MIM: 609600
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF396 PubMed Links

Gene Details

Gene
ZNF396
Gene Name
zinc finger protein 396
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322286.1 848 Missense Mutation GAA,GCA E239A NP_001309215.1
NM_001322290.1 848 Missense Mutation GAA,GCA E239A NP_001309219.1
NM_001322291.1 848 Intron NP_001309220.1
NM_145756.2 848 Missense Mutation GAA,GCA E239A NP_665699.1
XM_006722432.3 848 Missense Mutation GAA,GCA E239A XP_006722495.1
XM_017025687.1 848 Missense Mutation GAA,GCA E239A XP_016881176.1
XM_017025688.1 848 Missense Mutation GAA,GCA E239A XP_016881177.1
XM_017025689.1 848 Intron XP_016881178.1

View Full Product Details