Product Details

SNP ID
rs200378187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:77251150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCTCGCCACAAGAAGGCCTACG[C/T]GGTGTGCACCTTCGTCTTCGGCTAC
Phenotype
MIM: 600377
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALR1 PubMed Links

Gene Details

Gene
GALR1
Gene Name
galanin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001480.3 1099 Missense Mutation GCG,GTG A201V NP_001471.2
XM_017025691.1 1099 Missense Mutation GCG,GTG A201V XP_016881180.1

View Full Product Details