Product Details
- SNP ID
-
rs200243252
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:35253973 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAGGTCTCTCTCCAGTATGAATT[A/C]TCTGATGTCTAATCAGCTTTGAGCT
- Phenotype
-
MIM: 609601
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF397
PubMed Links
Gene Details
- Gene
- ZNF397
- Gene Name
- zinc finger protein 397
- Gene
- ZSCAN30
- Gene Name
- zinc finger and SCAN domain containing 30
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001112734.3 |
1379 |
Missense Mutation |
AGA,ATA |
R321I |
NP_001106205.1 |
NM_001166012.2 |
1379 |
Missense Mutation |
AGA,ATA |
R321I |
NP_001159484.1 |
NM_001288711.1 |
1379 |
Missense Mutation |
AGA,ATA |
R134I |
NP_001275640.1 |
XM_005258183.4 |
1379 |
Missense Mutation |
AGA,ATA |
R321I |
XP_005258240.1 |
XM_006722371.3 |
1379 |
Missense Mutation |
AGA,ATA |
R321I |
XP_006722434.1 |
XM_011525789.2 |
1379 |
Missense Mutation |
AGA,ATA |
R134I |
XP_011524091.1 |
XM_017025515.1 |
1379 |
Missense Mutation |
AGA,ATA |
R321I |
XP_016881004.1 |
XM_017025516.1 |
1379 |
Missense Mutation |
AGA,ATA |
R178I |
XP_016881005.1 |
XM_017025517.1 |
1379 |
Missense Mutation |
AGA,ATA |
R178I |
XP_016881006.1 |
XM_017025518.1 |
1379 |
Missense Mutation |
AGA,ATA |
R178I |
XP_016881007.1 |
XM_017025519.1 |
1379 |
Missense Mutation |
AGA,ATA |
R134I |
XP_016881008.1 |
XM_017025520.1 |
1379 |
Missense Mutation |
AGA,ATA |
R134I |
XP_016881009.1 |
XM_017025521.1 |
1379 |
Missense Mutation |
AGA,ATA |
R134I |
XP_016881010.1 |
XM_017025522.1 |
1379 |
Missense Mutation |
AGA,ATA |
R134I |
XP_016881011.1 |
View Full Product Details