Product Details

SNP ID
rs201095674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63637958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAGGTCATGCCTGAGAGGTCTG[A/C]ATCCCCATTGAAGATATTCACCATT
Phenotype
MIM: 600518
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SERPINB4 PubMed Links

Gene Details

Gene
SERPINB4
Gene Name
serpin family B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002974.3 1081 Missense Mutation GCA,TCA A312S NP_002965.1
NM_175041.1 1081 Missense Mutation GCA,TCA A291S NP_778206.1
XM_011526138.1 1081 Missense Mutation GCA,TCA A312S XP_011524440.1

View Full Product Details