Product Details

SNP ID
rs201239078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:580662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGGAAGGCACGGGGAAGATCA[A/G]CTTCAATGACTTCCTGGCCGTGATG
Phenotype
MIM: 603187
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CETN1 PubMed Links

Gene Details

Gene
CETN1
Gene Name
centrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004066.2 320 Missense Mutation AAC,AGC N85S NP_004057.1

View Full Product Details