Product Details

SNP ID
rs201123864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57436862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAAAACCGTGGAGTAAACTCAA[A/G]TCTGGCAGGGAGACCTTCCGCAGGA
Phenotype
MIM: 604894
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ONECUT2 PubMed Links

Gene Details

Gene
ONECUT2
Gene Name
one cut homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004852.2 1443 Silent Mutation AAA,AAG K382K NP_004843.2
XM_017026096.1 1443 Silent Mutation AAA,AAG K382K XP_016881585.1

View Full Product Details