Product Details

SNP ID
rs200197635
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57978979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCCACATTCAGTACATTTATA[C/T]GGTTTTGCTCCAGAATGAGTTCTTT
Phenotype
MIM: 613905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf18 PubMed Links

Gene Details

Gene
C19orf18
Gene Name
chromosome 19 open reading frame 18
There are no transcripts associated with this gene.

Gene
ZNF606
Gene Name
zinc finger protein 606
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025027.3 2179 Silent Mutation CCA,CCG P567P NP_079303.2
XM_005259276.1 2179 Silent Mutation CCA,CCG P567P XP_005259333.1

View Full Product Details