Product Details
- SNP ID
-
rs200069099
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:14151344 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCGGGCCCGCCCGCCTCTTCCTC[A/G]CCCCCGCCCCCTGGCACAGGTGGCA
- Phenotype
-
MIM: 616416
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRL1
PubMed Links
Gene Details
- Gene
- ADGRL1
- Gene Name
- adhesion G protein-coupled receptor L1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001008701.2 |
4363 |
Silent Mutation |
GGC,GGT |
G1318G |
NP_001008701.1 |
NM_014921.4 |
4363 |
Silent Mutation |
GGC,GGT |
G1313G |
NP_055736.2 |
XM_005259818.3 |
4363 |
Silent Mutation |
GGC,GGT |
G1337G |
XP_005259875.1 |
XM_011527796.2 |
4363 |
Silent Mutation |
GGC,GGT |
G1349G |
XP_011526098.1 |
XM_011527798.2 |
4363 |
Silent Mutation |
GGC,GGT |
G1344G |
XP_011526100.1 |
XM_011527801.2 |
4363 |
Silent Mutation |
GGC,GGT |
G1269G |
XP_011526103.1 |
XM_017026475.1 |
4363 |
Silent Mutation |
GGC,GGT |
G1350G |
XP_016881964.1 |
XM_017026476.1 |
4363 |
Silent Mutation |
GGC,GGT |
G1350G |
XP_016881965.1 |
XM_017026477.1 |
4363 |
Silent Mutation |
GGC,GGT |
G1343G |
XP_016881966.1 |
XM_017026478.1 |
4363 |
Silent Mutation |
GGC,GGT |
G1326G |
XP_016881967.1 |
XM_017026479.1 |
4363 |
Silent Mutation |
GGC,GGT |
G1320G |
XP_016881968.1 |
XM_017026480.1 |
4363 |
Silent Mutation |
GGC,GGT |
G1264G |
XP_016881969.1 |
XM_017026481.1 |
4363 |
Intron |
|
|
XP_016881970.1 |
- Gene
- LOC100507373
- Gene Name
- uncharacterized LOC100507373
There are no transcripts associated with this gene.
View Full Product Details