Product Details

SNP ID
rs199580200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5623331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGAGTGATCGATCTGCGGGCGGA[A/G]CTGAGGAAACGGAATGTGGACTCGA
Phenotype
MIM: 602895 MIM: 608066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAFB PubMed Links

Gene Details

Gene
SAFB
Gene Name
scaffold attachment factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201338.1 297 Silent Mutation GAA,GAG E42E NP_001188267.1
NM_001201339.1 297 Silent Mutation GAA,GAG E42E NP_001188268.1
NM_001201340.1 297 Silent Mutation GAA,GAG E42E NP_001188269.1
NM_001320571.1 297 UTR 5 NP_001307500.1
NM_001320572.1 297 Silent Mutation GAA,GAG E42E NP_001307501.1
NM_002967.3 297 Silent Mutation GAA,GAG E42E NP_002958.2
XM_017027114.1 297 UTR 5 XP_016882603.1
XM_017027115.1 297 UTR 5 XP_016882604.1
XM_017027116.1 297 UTR 5 XP_016882605.1
XM_017027117.1 297 UTR 5 XP_016882606.1
XM_017027118.1 297 UTR 5 XP_016882607.1
Gene
SAFB2
Gene Name
scaffold attachment factor B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014649.2 297 Intron NP_055464.1
XM_011528449.2 297 Intron XP_011526751.1
XM_011528450.2 297 Intron XP_011526752.1
XM_017027520.1 297 Intron XP_016883009.1
XM_017027521.1 297 Intron XP_016883010.1
XM_017027522.1 297 Intron XP_016883011.1

View Full Product Details