Product Details

SNP ID
rs200077528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33137719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGGCACGAGCACGCTGTGAGC[A/C]CCTGCCACTTCTGTGTGGATGACAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPATCH1 PubMed Links

Gene Details

Gene
GPATCH1
Gene Name
G-patch domain containing 1
There are no transcripts associated with this gene.

Gene
WDR88
Gene Name
WD repeat domain 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173479.3 391 Missense Mutation ACC,CCC T107P NP_775750.3
XM_011526449.2 391 Missense Mutation ACC,CCC T107P XP_011524751.1
XM_011526450.2 391 Missense Mutation ACC,CCC T107P XP_011524752.1
XM_017026282.1 391 Missense Mutation ACC,CCC T107P XP_016881771.1
XM_017026283.1 391 Missense Mutation ACC,CCC T107P XP_016881772.1

View Full Product Details