Product Details

SNP ID
rs200400365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55644635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCCTCCGTTGAGACCATGGAG[A/G]GCCCTCCCCGTCGGACTTGCCGCTC
Phenotype
MIM: 613478 MIM: 191318
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC106 PubMed Links

Gene Details

Gene
CCDC106
Gene Name
coiled-coil domain containing 106
There are no transcripts associated with this gene.

Gene
LOC107983998
Gene Name
uncharacterized LOC107983998
There are no transcripts associated with this gene.

Gene
U2AF2
Gene Name
U2 small nuclear RNA auxiliary factor 2
There are no transcripts associated with this gene.

Gene
ZNF580
Gene Name
zinc finger protein 580
There are no transcripts associated with this gene.

Gene
ZNF581
Gene Name
zinc finger protein 581
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016535.3 238 Missense Mutation AGC,GGC S22G NP_057619.1
XM_006723240.3 238 Missense Mutation AGC,GGC S22G XP_006723303.1
XM_017026867.1 238 Missense Mutation AGC,GGC S22G XP_016882356.1

View Full Product Details