Product Details

SNP ID
rs200016199
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55948126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACCATGCCCATCACCTGGGACC[A/G]GGTCGAGACAGCCAGCTGGGCAGAG
Phenotype
MIM: 609659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLRP8 PubMed Links

Gene Details

Gene
NLRP8
Gene Name
NLR family pyrin domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317000.1 295 Missense Mutation CAG,CGG Q75R NP_001303929.1
NM_176811.2 295 Missense Mutation CAG,CGG Q75R NP_789781.2

View Full Product Details