Product Details

SNP ID
rs200444143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1398965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTTGGACTTCATCAGCTCCCCC[A/C]AGGAGGTGAGGTTGCAGTAGGTGAG
Phenotype
MIM: 607430 MIM: 601240 MIM: 601825
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DAZAP1 PubMed Links

Gene Details

Gene
DAZAP1
Gene Name
DAZ associated protein 1
There are no transcripts associated with this gene.

Gene
GAMT
Gene Name
guanidinoacetate N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000156.5 615 Missense Mutation NP_000147.1
NM_138924.2 615 Missense Mutation NP_620279.1
Gene
NDUFS7
Gene Name
NADH:ubiquinone oxidoreductase core subunit S7
There are no transcripts associated with this gene.

View Full Product Details