Product Details

SNP ID
rs199844955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7075080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGCTGTCGGAGTCACAGGATGGC[C/G]GCTGTCGTCCTGCCCCCAACTGCCG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF557 PubMed Links

Gene Details

Gene
ZNF557
Gene Name
zinc finger protein 557
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044387.1 491 Silent Mutation GCC,GCG A2A NP_001037852.1
NM_001044388.1 491 Silent Mutation GCC,GCG A2A NP_001037853.1
NM_024341.2 491 Silent Mutation GCC,GCG A2A NP_077317.2

View Full Product Details