Product Details

SNP ID
rs202169240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50724046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCAACGCCAACCCCATCCTCC[C/G]GCCCCAGCCCCTCTCCCACCCCTGA
Phenotype
MIM: 604713 MIM: 604999
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLEC11A PubMed Links

Gene Details

Gene
CLEC11A
Gene Name
C-type lectin domain family 11 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002975.2 467 Missense Mutation CGC,GGC R97G NP_002966.1
Gene
SHANK1
Gene Name
SH3 and multiple ankyrin repeat domains 1
There are no transcripts associated with this gene.

View Full Product Details