Product Details
- SNP ID
-
rs200920864
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:43718978 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCGCTATGACTTCTTCCTGCTGGT[A/C]TCCCCCCGCCGCTGCGGGGCCGTCG
- Phenotype
-
MIM: 613176
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IRGC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs346049] are located under a probe and SNP(s) [rs11555891] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IRGC
- Gene Name
- immunity related GTPase cinema
- Gene
- SMG9
- Gene Name
- SMG9, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.
View Full Product Details