Product Details

SNP ID
rs200681860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18532184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTAGGTGGTCAGTTCCTGGCAGC[A/G]ATGACCACAGAGAGTGCCACACCCC
Phenotype
MIM: 604840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FKBP8 PubMed Links

Gene Details

Gene
FKBP8
Gene Name
FK506 binding protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308373.1 1421 Silent Mutation ATC,ATT I408I NP_001295302.1
NM_012181.4 1421 Silent Mutation ATC,ATT I409I NP_036313.3
XM_011527863.1 1421 Silent Mutation ATC,ATT I409I XP_011526165.1
XM_011527864.1 1421 UTR 3 XP_011526166.1

View Full Product Details