Product Details

SNP ID
rs202240553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48603822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCGGGCAGGAGGCCGCGTACAGC[A/G]TCCGGAACTCAAGGCTGAAGGCGTC
Phenotype
MIM: 609932 MIM: 604125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM83E PubMed Links

Gene Details

Gene
FAM83E
Gene Name
family with sequence similarity 83 member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017708.3 913 Missense Mutation ACG,ATG T283M NP_060178.2
XM_005259019.1 913 Missense Mutation ACG,ATG T283M XP_005259076.1
XM_011527049.2 913 Intron XP_011525351.1
Gene
SPACA4
Gene Name
sperm acrosome associated 4
There are no transcripts associated with this gene.

Gene
SULT2B1
Gene Name
sulfotransferase family 2B member 1
There are no transcripts associated with this gene.

View Full Product Details