Product Details

SNP ID
rs200619245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43535560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCTATGTCACGACCCCCCAACCG[A/C]GCCCGGCAGCCAGGCGACTGCCTGG
Phenotype
MIM: 608451 MIM: 194360
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ETHE1 PubMed Links

Gene Details

Gene
ETHE1
Gene Name
ETHE1, persulfide dioxygenase
There are no transcripts associated with this gene.

Gene
XRCC1
Gene Name
X-ray repair cross complementing 1
There are no transcripts associated with this gene.

Gene
ZNF575
Gene Name
zinc finger protein 575
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174945.2 881 Missense Mutation GAG,GCG E204A NP_777605.1
XM_005258783.3 881 Missense Mutation GAG,GCG E204A XP_005258840.1
XM_011526793.2 881 Missense Mutation GAG,GCG E204A XP_011525095.1

View Full Product Details