Product Details

SNP ID
rs201168463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52434063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGACGTGATGTTAGAGAACTAC[A/G]GGAACCTGGTCTCCCTAGGTGAGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF534 PubMed Links

Gene Details

Gene
ZNF534
Gene Name
zinc finger protein 534
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143938.1 185 Missense Mutation AGG,GGG R42G NP_001137410.1
NM_001143939.1 185 Missense Mutation AGG,GGG R42G NP_001137411.1
NM_001291368.1 185 Missense Mutation AGG,GGG R42G NP_001278297.1
NM_001291369.1 185 Missense Mutation AGG,GGG R42G NP_001278298.1

View Full Product Details