Product Details

SNP ID
rs201551006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9115193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGGATGTTGTTGATGAGGGTGT[C/T]AGAACAGGCGAGCTTGATGACCTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR7G1 PubMed Links

Gene Details

Gene
OR7G1
Gene Name
olfactory receptor family 7 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005192.2 571 Missense Mutation AAC,GAC N191D NP_001005192.2

View Full Product Details