Product Details

SNP ID
rs201000725
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4546263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCTGGCCCCGGACACGTCCTGC[G/T]CAAAGGCGGCTCTAATGGGGAGAGG
Phenotype
MIM: 611289 MIM: 608873
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
LRG1 PubMed Links

Gene Details

Gene
LRG1
Gene Name
leucine rich alpha-2-glycoprotein 1
There are no transcripts associated with this gene.

Gene
SEMA6B
Gene Name
semaphorin 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032108.3 1953 Missense Mutation GAG,GCG E564A NP_115484.2
XM_011527639.2 1953 Missense Mutation GAG,GCG E570A XP_011525941.1
XM_011527640.2 1953 Missense Mutation GAG,GCG E570A XP_011525942.1
XM_011527641.1 1953 Missense Mutation AGC,CGC S568R XP_011525943.1

View Full Product Details