Product Details

SNP ID
rs201866900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51481559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAATTTCCCCAGACGGCAAGAC[C/T]CTCGTCATCCTCAGGGTCAGCCGCT
Phenotype
MIM: 606094
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEACAM18 PubMed Links

Gene Details

Gene
CEACAM18
Gene Name
carcinoembryonic antigen related cell adhesion molecule 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278392.1 567 Silent Mutation ACC,ACT T189T NP_001265321.1
XM_011527258.2 567 Silent Mutation ACC,ACT T189T XP_011525560.1
Gene
SIGLEC12
Gene Name
sialic acid binding Ig like lectin 12 (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details