Product Details

SNP ID
rs200937372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12701761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAGCTGCCCCAACCTGCACAAC[A/G]CCCCCCGGGTTGACACCGATCATCA
Phenotype
MIM: 609074 MIM: 603002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXW9 PubMed Links

Gene Details

Gene
FBXW9
Gene Name
F-box and WD repeat domain containing 9
There are no transcripts associated with this gene.

Gene
SNORD135
Gene Name
small nucleolar RNA, C/D box 135
There are no transcripts associated with this gene.

Gene
SNORD41
Gene Name
small nucleolar RNA, C/D box 41
There are no transcripts associated with this gene.

Gene
TNPO2
Gene Name
transportin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136195.1 2643 Silent Mutation GGC,GGT G824G NP_001129667.1
NM_001136196.1 2643 Silent Mutation GGC,GGT G834G NP_001129668.1
NM_013433.4 2643 Silent Mutation GGC,GGT G824G NP_038461.2

View Full Product Details